siRNA Details
Gene Symbol: Azin1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301702.1 | NP_001288631.1 | 6 | 1184 | ||
NM_022585.2 | NP_072107.2 | 7 | 1342 | |||
XM_006241568.2 | XP_006241630.1 | 7 | 1315 | |||
XM_006241569.2 | XP_006241631.1 | 6 | 1179 | |||
XM_008765457.1 | XP_008763679.1 | 6 | 1179 | |||
GenBank mRNA | BC061550.1 | - | 4 | 580 | 6918922 | |
D50734.1 | - | 6 | 977 | |||
D89983.1 | - | 7 | 1290 |
