siRNA Details
Gene Symbol: Azin1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301702.1 | NP_001288631.1 | 4 | 771 | ||
NM_022585.2 | NP_072107.2 | 5 | 929 | |||
XM_006241568.2 | XP_006241630.1 | 5 | 902 | |||
XM_006241569.2 | XP_006241631.1 | 4 | 766 | |||
XM_008765457.1 | XP_008763679.1 | 4 | 766 | |||
GenBank mRNA | BC061550.1 | - | 2 | 167 | 6918922 | |
D50734.1 | - | 4 | 564 | |||
D89983.1 | - | 5 | 877 |
