siRNA Details
Gene Symbol: Ncan | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031653.2 | NP_113841.2 | 3 | 221 | ||
GenBank mRNA | M97161.1 | - | 3 | 221 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ncan | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031653.2 | NP_113841.2 | 3 | 221 | ||
GenBank mRNA | M97161.1 | - | 3 | 221 |