siRNA Details
| Gene Symbol: Milr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021585.1 | NP_067596.1 | 4 | 511 | ||
| GenBank mRNA | U39546.1 | - | 4 | 511 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Milr1| Gene Symbol: Milr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021585.1 | NP_067596.1 | 4 | 511 | ||
| GenBank mRNA | U39546.1 | - | 4 | 511 | ||