siRNA Details
| Gene Symbol: Trdn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021666.2 | NP_067698.1 | 2049 | |||
| GenBank mRNA | AJ243304.2 | - | 2049 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Trdn| Gene Symbol: Trdn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021666.2 | NP_067698.1 | 2049 | |||
| GenBank mRNA | AJ243304.2 | - | 2049 | |||