siRNA Details
Gene Symbol: Epn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033914.1 | NP_001029086.1 | 7, 8 | 1599 | ||
NM_021852.2 | NP_068624.2 | 6, 7 | 1428 | |||
XM_006246521.2 | XP_006246583.1 | 7, 8 | 1383 | |||
XM_006246522.2 | XP_006246584.1 | 8, 9 | 1681 | |||
XM_006246523.2 | XP_006246585.1 | 6, 7 | 1212 | |||
GenBank mRNA | AF096269.1 | - | 5, 6 | 1169 | ||
BC094524.1 | - | 7, 8 | 1578 | 7318094 | ||
FQ211574.1 | - | 8, 9 | 1681 | |||
FQ211939.1 | - | 7, 8 | 1623 |
