siRNA Details
| Gene Symbol: Syt10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031666.1 | NP_113854.1 | 2, 3 | 503 | ||
| GenBank mRNA | AF375463.1 | - | 2, 3 | 503 | ||
| DQ437524.1 | - | 2, 3 | 503 | |||
| U85513.1 | - | 2, 3 | 635 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Syt10| Gene Symbol: Syt10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031666.1 | NP_113854.1 | 2, 3 | 503 | ||
| GenBank mRNA | AF375463.1 | - | 2, 3 | 503 | ||
| DQ437524.1 | - | 2, 3 | 503 | |||
| U85513.1 | - | 2, 3 | 635 | |||