siRNA Details
| Gene Symbol: Syt11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031667.2 | NP_113855.2 | 2 | 365 | ||
| XM_006232748.1 | XP_006232810.1 | 2 | 357 | |||
| GenBank mRNA | AF000423.1 | - | 2 | 406 | ||
| AF375465.1 | - | 2 | 164 | |||
| BC094518.1 | - | 2 | 365 | 7315033 | ||

Syt11
Genomic Map