siRNA Details
| Gene Symbol: Lif | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022196.2 | NP_071532.2 | 3 | 357 | ||
| XM_006251364.2 | XP_006251426.1 | 4 | 492 | |||
| XM_008770359.1 | XP_008768581.1 | 3 | 1062 | |||
| GenBank mRNA | AB010275.1 | - | 3 | 357 | ||
| M32748.1 | - | 3 | 357 | |||

Lif
Genomic Map