siRNA Details
| Gene Symbol: Ppp2r2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022209.3 | NP_071545.2 | 7 | 1059 | ||
| GenBank mRNA | AY251277.1 | - | 7 | 1011 | ||
| BC078834.1 | - | 10 | 1268 | 7114294 | ||
| D38260.1 | - | 7 | 854 | |||
| FQ212581.1 | - | 5 | 579 | |||

Ppp2r2b
Genomic Map