siRNA Details
| Gene Symbol: Fgf14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022223.2 | NP_071559.2 | 2 | 240 | ||
| GenBank mRNA | AB008908.1 | - | 255 | |||
| AB079500.1 | - | 2 | 240 | |||
| AF348523.1 | - | 134 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fgf14| Gene Symbol: Fgf14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022223.2 | NP_071559.2 | 2 | 240 | ||
| GenBank mRNA | AB008908.1 | - | 255 | |||
| AB079500.1 | - | 2 | 240 | |||
| AF348523.1 | - | 134 | ||||