siRNA Details
Gene Symbol: Pter | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022224.2 | NP_071560.2 | 2 | 308 | ||
XM_006254290.2 | XP_006254352.2 | 2 | 3577 | |||
XM_006254291.2 | XP_006254353.1 | 2 | 395 | |||
GenBank mRNA | BC078833.1 | - | 2 | 308 | 7110694 |
