siRNA Details
| Gene Symbol: Tdo2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022403.2 | NP_071798.1 | 12 | 1226 | ||
| GenBank mRNA | BC089802.1 | - | 12 | 1222 | 7370033 | |
| M55167.1 | - | 12 | 1226 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tdo2| Gene Symbol: Tdo2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022403.2 | NP_071798.1 | 12 | 1226 | ||
| GenBank mRNA | BC089802.1 | - | 12 | 1222 | 7370033 | |
| M55167.1 | - | 12 | 1226 | |||