siRNA Details
| Gene Symbol: Sult1b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022513.2 | NP_071958.1 | 7 | 912 | ||
| XM_008770056.1 | XP_008768278.1 | 7 | 786 | |||
| GenBank mRNA | D89375.1 | - | 7 | 863 | ||
| FQ209476.1 | - | 7 | 880 | |||
| FQ209486.1 | - | 7 | 947 | |||
| FQ209579.1 | - | 7 | 967 | |||
| FQ210801.1 | - | 7 | 954 | |||
| FQ213902.1 | - | 2 | 176 | |||
| FQ218295.1 | - | 7 | 925 | |||
| U38419.1 | - | 7 | 915 | |||

Sult1b1
Genomic Map