siRNA Details
| Gene Symbol: Arf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022518.3 | NP_071963.1 | 5 | 574 | ||
| XM_006246525.2 | XP_006246587.1 | 5 | 621 | |||
| XM_008767870.1 | XP_008766092.1 | 6 | 666 | |||
| GenBank mRNA | BC061552.1 | - | 5 | 590 | 6889332 | |
| FQ217649.1 | - | 2 | 365 | |||
| L12380.1 | - | 5 | 574 | |||

Arf1
Genomic Map