siRNA Details
| Gene Symbol: Gstm5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172038.1 | NP_742035.1 | 5, 6 | 379 | ||
| GenBank mRNA | U86635.1 | - | 5, 6 | 379 | ||
| Gene Symbol: LOC100912430 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003749378.3 | XP_003749426.1 | 5, 6 | 515 | ||

Gstm5
Genomic Map