siRNA Details
| Gene Symbol: Aldh1l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022547.1 | NP_071992.1 | 12 | 1763 | ||
| GenBank mRNA | BC089101.1 | - | 12 | 1772 | 7306458 | |
| M59861.1 | - | 12 | 1763 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Aldh1l1| Gene Symbol: Aldh1l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022547.1 | NP_071992.1 | 12 | 1763 | ||
| GenBank mRNA | BC089101.1 | - | 12 | 1772 | 7306458 | |
| M59861.1 | - | 12 | 1763 | |||