siRNA Details
| Gene Symbol: Pex14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172063.1 | NP_742060.1 | 4 | 201 | ||
| GenBank mRNA | AB017544.1 | - | 4 | 201 | ||
| BC081708.1 | - | 4 | 190 | 7191447 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pex14| Gene Symbol: Pex14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172063.1 | NP_742060.1 | 4 | 201 | ||
| GenBank mRNA | AB017544.1 | - | 4 | 201 | ||
| BC081708.1 | - | 4 | 190 | 7191447 | ||