siRNA Details
| Gene Symbol: Decr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_171996.3 | NP_741993.1 | 5 | 460 | ||
| XM_006246024.2 | XP_006246086.1 | 6 | 660 | |||
| GenBank mRNA | AF044574.1 | - | 5 | 382 | ||
| BC070959.1 | - | 5 | 461 | 7097883 | ||
| FQ218511.1 | - | 5 | 435 | |||

Decr2
Genomic Map