siRNA Details
| Gene Symbol: Csnk1d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139060.3 | NP_620691.2 | 4 | 850 | ||
| XM_008768511.1 | XP_008766733.1 | 4 | 855 | |||
| GenBank mRNA | AB063114.1 | - | 4 | 842 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Csnk1d| Gene Symbol: Csnk1d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139060.3 | NP_620691.2 | 4 | 850 | ||
| XM_008768511.1 | XP_008766733.1 | 4 | 855 | |||
| GenBank mRNA | AB063114.1 | - | 4 | 842 | ||