siRNA Details
| Gene Symbol: Thop1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172075.2 | NP_742072.2 | 3 | 460 | ||
| XM_006240995.2 | XP_006241057.1 | 3 | 480 | |||
| GenBank mRNA | BC081706.1 | - | 3 | 387 | 7111806 | |
| M61142.1 | - | 3 | 383 | |||

Thop1
Genomic Map