siRNA Details
| Gene Symbol: Cnbp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022598.1 | NP_072120.1 | 4 | 452 | ||
| XM_006236854.2 | XP_006236916.1 | 4 | 447 | |||
| GenBank mRNA | AB158421.1 | - | 3 | 287 | ||
| AB158422.1 | - | 3 | 287 | |||
| AF242550.1 | - | 4 | 338 | |||
| BC062225.1 | - | 4 | 436 | 6889682 | ||
| D45254.1 | - | 4 | 452 | |||

Cnbp
Genomic Map