siRNA Details
Gene Symbol: B3gat2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022609.2 | NP_072131.1 | 4 | 1599 | ||
GenBank mRNA | AB010441.1 | - | 4 | 1599 | ||
AF106624.1 | - | 4 | 1594 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: B3gat2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022609.2 | NP_072131.1 | 4 | 1599 | ||
GenBank mRNA | AB010441.1 | - | 4 | 1599 | ||
AF106624.1 | - | 4 | 1594 |