siRNA Details
| Gene Symbol: Tmprss11d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033652.1 | NP_001028824.1 | 10 | 1434 | ||
| NM_022630.1 | NP_072152.1 | 5 | 805 | |||
| GenBank mRNA | AF198087.1 | - | 5 | 805 | ||
| AF453776.1 | - | 10 | 1434 | |||

Tmprss11d
Genomic Map