siRNA Details
| Gene Symbol: Wnt5a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022631.1 | NP_072153.1 | 3 | 508 | ||
| XM_006252664.2 | XP_006252726.1 | 3 | 375 | |||
| XM_006252665.2 | XP_006252727.1 | 3 | 337 | |||
| XM_008771007.1 | XP_008769229.1 | 3 | 1213 | |||
| GenBank mRNA | AB244721.1 | - | 2 | 271 | ||
| AF188333.1 | - | 3 | 508 | |||
| AY819646.1 | - | 2 | 271 | |||

Wnt5a
Genomic Map