siRNA Details
| Gene Symbol: Bid | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022684.1 | NP_073175.1 | 5 | 706 | ||
| GenBank mRNA | AF259503.1 | - | 5 | 706 | ||
| D89804.1 | - | 662 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Bid| Gene Symbol: Bid | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022684.1 | NP_073175.1 | 5 | 706 | ||
| GenBank mRNA | AF259503.1 | - | 5 | 706 | ||
| D89804.1 | - | 662 | ||||