siRNA Details
| Gene Symbol: Bad | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022698.1 | NP_073189.1 | 2 | 133 | ||
| XM_006230896.2 | XP_006230958.1 | 2 | 838 | |||
| XM_006230897.2 | XP_006230959.1 | 1 | 16 | |||
| XM_006230898.2 | XP_006230960.1 | 2 | 31 | |||
| GenBank mRNA | AF003523.1 | - | 2 | 133 | ||
| AF031227.1 | - | 2 | 171 | |||
| AF279910.1 | - | 2 | 133 | |||
| AF279911.1 | - | 2 | 133 | |||
| BC061728.1 | - | 2 | 378 | 5598682 | ||

Bad
Genomic Map