siRNA Details
| Gene Symbol: Bad | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022698.1 | NP_073189.1 | 3 | 463 | ||
| XM_006230896.2 | XP_006230958.1 | 3 | 1168 | |||
| XM_006230897.2 | XP_006230959.1 | 2 | 346 | |||
| XM_006230898.2 | XP_006230960.1 | 3 | 361 | |||
| GenBank mRNA | AF031227.1 | - | 3 | 501 | ||
| AF279910.1 | - | 3 | 463 | |||
| AF279911.1 | - | 3 | 463 | |||

Bad
Genomic Map