siRNA Details
| Gene Symbol: Arl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031711.1 | NP_113899.1 | 2, 3 | 214 | ||
| XM_006230899.1 | XP_006230961.1 | 2, 3 | 263 | |||
| GenBank mRNA | FQ213929.1 | - | 2, 3 | 222 | ||
| Y12708.1 | - | 2, 3 | 214 | |||

Arl2
Genomic Map