siRNA Details
Gene Symbol: Rab2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031718.1 | NP_113906.1 | 1 | 20 | ||
GenBank mRNA | FQ229541.1 | - | 1 | 222 | ||
J02999.1 | - | 1 | 20 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Rab2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031718.1 | NP_113906.1 | 1 | 20 | ||
GenBank mRNA | FQ229541.1 | - | 1 | 222 | ||
J02999.1 | - | 1 | 20 |