siRNA Details
Gene Symbol: Dio2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031720.3 | NP_113908.3 | 1 | 602 | ||
GenBank mRNA | U53505.1 | - | 1 | 602 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Dio2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031720.3 | NP_113908.3 | 1 | 602 | ||
GenBank mRNA | U53505.1 | - | 1 | 602 |