siRNA Details
Gene Symbol: Prol1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133513.1 | NP_598197.1 | 3 | 499 | ||
GenBank mRNA | U03407.1 | - | 3 | 499 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Prol1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133513.1 | NP_598197.1 | 3 | 499 | ||
GenBank mRNA | U03407.1 | - | 3 | 499 |