siRNA Details
| Gene Symbol: Csnk1g2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033870.1 | NP_001029042.1 | 9 | 1426 | ||
| NM_023102.2 | NP_075590.2 | 9 | 1423 | |||
| XM_006241000.2 | XP_006241062.1 | 8 | 2801 | |||
| XM_006241001.2 | XP_006241063.1 | 8 | 1509 | |||
| XM_006241002.2 | XP_006241064.1 | 9 | 2721 | |||
| XM_006241003.2 | XP_006241065.1 | 8 | 2151 | |||
| XM_008765130.1 | XP_008763352.1 | 8 | 1509 | |||
| GenBank mRNA | BC072533.1 | - | 9 | 1426 | 7098992 | |
| BC099758.1 | - | 3 | 285 | |||
| U22297.1 | - | 8 | 1157 | |||

Csnk1g2
Genomic Map