siRNA Details
| Gene Symbol: Csnk1g2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033870.1 | NP_001029042.1 | 6 | 1009 | ||
| NM_023102.2 | NP_075590.2 | 6 | 1006 | |||
| XM_006241000.2 | XP_006241062.1 | 5 | 2384 | |||
| XM_006241001.2 | XP_006241063.1 | 5 | 1092 | |||
| XM_006241002.2 | XP_006241064.1 | 6 | 2304 | |||
| XM_006241003.2 | XP_006241065.1 | 5 | 1734 | |||
| XM_008765130.1 | XP_008763352.1 | 5 | 1092 | |||
| GenBank mRNA | BC072533.1 | - | 6 | 1009 | 7098992 | |
| U22297.1 | - | 5 | 740 | |||

Csnk1g2
Genomic Map