siRNA Details
| Gene Symbol: Kcnmb4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023960.2 | NP_076450.1 | 1, 2 | 363 | ||
| GenBank mRNA | AB050637.1 | - | 1, 2 | 345 | ||
| AY028605.1 | - | 1, 2 | 363 | |||
| BC161829.1 | - | 1, 2 | 543 | 9033053 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kcnmb4| Gene Symbol: Kcnmb4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023960.2 | NP_076450.1 | 1, 2 | 363 | ||
| GenBank mRNA | AB050637.1 | - | 1, 2 | 345 | ||
| AY028605.1 | - | 1, 2 | 363 | |||
| BC161829.1 | - | 1, 2 | 543 | 9033053 | ||