siRNA Details
| Gene Symbol: Kiss1r | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001301151.1 | NP_001288080.1 | 3 | 567 | ||
| NM_023992.2 | NP_076482.2 | 3 | 567 | |||
| GenBank mRNA | AB051066.1 | - | 3 | 393 | ||
| AF115516.1 | - | 3 | 393 | |||
More Information
Set Membership: |
|

Kiss1r
Genomic Map