siRNA Details
| Gene Symbol: Arf5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024149.1 | NP_077063.1 | 3 | 310 | ||
| GenBank mRNA | BC087692.1 | - | 3 | 272 | 7310424 | |
| L12384.1 | - | 3 | 310 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Arf5| Gene Symbol: Arf5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024149.1 | NP_077063.1 | 3 | 310 | ||
| GenBank mRNA | BC087692.1 | - | 3 | 272 | 7310424 | |
| L12384.1 | - | 3 | 310 | |||