siRNA Details
| Gene Symbol: Arf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024150.2 | NP_077064.1 | 4 | 392 | ||
| XM_006247543.2 | XP_006247605.1 | 4 | 490 | |||
| GenBank mRNA | BC098915.1 | - | 4 | 392 | 7461110 | |
| L12381.1 | - | 3 | 439 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Arf2| Gene Symbol: Arf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024150.2 | NP_077064.1 | 4 | 392 | ||
| XM_006247543.2 | XP_006247605.1 | 4 | 490 | |||
| GenBank mRNA | BC098915.1 | - | 4 | 392 | 7461110 | |
| L12381.1 | - | 3 | 439 | |||