siRNA Details
| Gene Symbol: Arf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024151.1 | NP_077065.1 | 3 | 406 | ||
| GenBank mRNA | BC063167.1 | - | 3 | 364 | 6921397 | |
| FQ213125.1 | - | 3 | 389 | |||
| FQ213804.1 | - | 3 | 403 | |||
| FQ234083.1 | - | 3 | 364 | |||
| FQ234248.1 | - | 3 | 476 | |||
| L12383.1 | - | 3 | 406 | |||

Arf4
Genomic Map