siRNA Details
| Gene Symbol: Asic1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024154.2 | NP_077068.1 | 2 | 426 | ||
| XM_006257381.2 | XP_006257443.1 | 2 | 711 | |||
| XM_008765794.1 | XP_008764016.1 | 2 | 711 | |||
| XM_008765795.1 | XP_008764017.1 | 2 | 364 | |||
| XM_008765796.1 | XP_008764018.1 | 2 | 711 | |||

Asic1
Genomic Map