siRNA Details
| Gene Symbol: Mtpn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024374.4 | NP_077350.1 | 2 | 240 | ||
| GenBank mRNA | AY951952.1 | - | 247 | |||
| BC088136.1 | - | 2 | 240 | 7377576 | ||
| D26179.1 | - | 2 | 295 | |||
| FQ228349.1 | - | 361 | ||||
| U21661.1 | - | 2 | 319 | |||

Mtpn
Genomic Map