siRNA Details
Gene Symbol: Mtpn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024374.4 | NP_077350.1 | 2 | 291 | ||
GenBank mRNA | AY951952.1 | - | 298 | |||
BC088136.1 | - | 2 | 291 | 7377576 | ||
D26179.1 | - | 2 | 346 | |||
FQ228349.1 | - | 412 | ||||
U21661.1 | - | 2 | 370 |
