siRNA Details
Gene Symbol: Hhex | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024385.1 | NP_077361.1 | 4 | 759 | ||
GenBank mRNA | BC088135.1 | - | 4 | 764 | 7366155 | |
D86383.1 | - | 4 | 759 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Hhex | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024385.1 | NP_077361.1 | 4 | 759 | ||
GenBank mRNA | BC088135.1 | - | 4 | 764 | 7366155 | |
D86383.1 | - | 4 | 759 |