siRNA Details
| Gene Symbol: Cdk5rap3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024488.1 | NP_077814.1 | 7 | 612 | ||
| GenBank mRNA | AF177476.1 | - | 7 | 612 | ||
| BC081793.1 | - | 6 | 534 | 7190258 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cdk5rap3| Gene Symbol: Cdk5rap3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024488.1 | NP_077814.1 | 7 | 612 | ||
| GenBank mRNA | AF177476.1 | - | 7 | 612 | ||
| BC081793.1 | - | 6 | 534 | 7190258 | ||