siRNA Details
| Gene Symbol: Cyb5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030586.2 | NP_085075.1 | 5 | 393 | ||
| GenBank mRNA | BC072535.1 | - | 5 | 393 | 7103272 | |
| X96392.1 | - | 5 | 261 | |||
| Y12517.1 | - | 5 | 438 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cyb5b| Gene Symbol: Cyb5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030586.2 | NP_085075.1 | 5 | 393 | ||
| GenBank mRNA | BC072535.1 | - | 5 | 393 | 7103272 | |
| X96392.1 | - | 5 | 261 | |||
| Y12517.1 | - | 5 | 438 | |||