siRNA Details
| Gene Symbol: Slco1a5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030838.1 | NP_110465.1 | 10 | 1431 | ||
| XM_006237642.2 | XP_006237704.1 | 12 | 1646 | |||
| XM_006237643.2 | XP_006237705.1 | 12 | 1666 | |||
| XM_006237646.2 | XP_006237708.1 | 10 | 1447 | |||
| XM_006237648.2 | XP_006237710.1 | 7 | 1859 | |||
| XM_008763432.1 | XP_008761654.1 | 11 | 3015 | |||
| GenBank mRNA | AF041105.1 | - | 9 | 1194 | ||
| AF083469.1 | - | 10 | 1431 | |||
| FQ211722.1 | - | 12 | 1661 | |||

Slco1a5
Genomic Map