siRNA Details
| Gene Symbol: Lig1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024268.2 | NP_001019439.1 | 28 | 2848 | ||
| GenBank mRNA | AF276774.1 | - | 28 | 2878 | ||
| BC093604.1 | - | 28 | 2832 | 7317599 | ||
| BE104054.1 | - | 1 | 218 | |||
| Gene Symbol: LOC100911727 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008759094.1 | XP_008757316.1 | 24 | 2517 | ||

Lig1
Genomic Map