siRNA Details
| Gene Symbol: Smad7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030858.1 | NP_110485.1 | 3 | 2110 | ||
| XM_006254959.2 | XP_006255021.1 | 3 | 704 | |||
| GenBank mRNA | AF042499.1 | - | 3 | 832 | ||
| AF159626.1 | - | 3 | 2110 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Smad7| Gene Symbol: Smad7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030858.1 | NP_110485.1 | 3 | 2110 | ||
| XM_006254959.2 | XP_006255021.1 | 3 | 704 | |||
| GenBank mRNA | AF042499.1 | - | 3 | 832 | ||
| AF159626.1 | - | 3 | 2110 | |||