siRNA Details
| Gene Symbol: Scn1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030875.1 | NP_110502.1 | 22 | 4417 | ||
| XM_008761923.1 | XP_008760145.1 | 22 | 4437 | |||
| XM_008761924.1 | XP_008760146.1 | 22 | 4345 | |||
| XM_008761925.1 | XP_008760147.1 | 22 | 4253 | |||
| XM_008761926.1 | XP_008760148.1 | 22 | 4436 | |||
| XM_008761927.1 | XP_008760149.1 | 22 | 4403 | |||
| XM_008761928.1 | XP_008760150.1 | 22 | 4401 | |||
| GenBank mRNA | M22253.1 | - | 22 | 4417 | ||
| X03638.1 | - | 22 | 4417 | |||

Scn1a
Genomic Map