siRNA Details
| Gene Symbol: Amd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031011.3 | NP_112273.3 | 6 | 836 | ||
| GenBank mRNA | AY724509.1 | - | 6 | 519 | ||
| BC061532.1 | - | 6 | 839 | 6921099 | ||
| FM031711.1 | - | 6 | 483 | |||
| FM123317.1 | - | 1 | 33 | |||
| FQ232080.1 | - | 6 | 656 | |||
| FQ234346.1 | - | 6 | 659 | |||
| FQ234398.1 | - | 1 | 1005 | |||
| M34464.1 | - | 6 | 782 | |||

Amd1
Genomic Map